5-Amino-2-chloro-N-cyclohexylbenzamide

Supplier: Matrix Scientific

024291-500MG
101817-802EA 306.15 USD
101817-802
5-Amino-2-chloro-N-cyclohexylbenzamide
5-Amino-2-chloro-N-cyclohexylbenzamide

Hazard: Irritant

Formula: C₁₃H₁₇ClN₂O
MDL Number: MFCD08728065
CAS Number: 920122-73-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR