2-Amino-N-ethyl-N-phenylbenzamide

Supplier: Matrix Scientific

024286-500MG
101817-794EA 180 USD
101817-794
2-Amino-N-ethyl-N-phenylbenzamide
2-Amino-N-ethyl-N-phenylbenzamide

Hazard: Irritant

Formula: C₁₅H₁₆N₂O
MDL Number: MFCD08728061
CAS Number: 29094-86-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR