N-(3-Amino-4-chlorophenyl)butanamide

Supplier: Matrix Scientific

024285-500MG
101817-792EA 306.15 USD
101817-792
N-(3-Amino-4-chlorophenyl)butanamide
N-(3-Amino-4-chlorophenyl)butanamide

Hazard: Irritant

Formula: C₁₀H₁₃ClN₂O
MDL Number: MFCD03723773
CAS Number: 637316-74-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR