2-Amino-N-sec-butylbenzamide

Supplier: Matrix Scientific

024277-500MG
101817-776EA 37.95 USD
101817-776
2-Amino-N-sec-butylbenzamide
2-Amino-N-sec-butylbenzamide

Hazard: Irritant

Formula: C₁₁H₁₆N₂O
MW: 192.26
MDL Number: MFCD00116303
CAS Number: 30391-87-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR