4-Hydroxybenzaldehyde oxime

Supplier: Matrix Scientific

024273-500MG
101817-768EA 306.15 USD
101817-768
4-Hydroxybenzaldehyde oxime
4-Hydroxybenzaldehyde oxime

Hazard: Irritant

Formula: C₇H₇NO₂
MDL Number: MFCD00019964
CAS Number: 60221-52-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR