2-[2-Bromo-4-(sec-butyl)phenoxy]acetic acid

Supplier: Matrix Scientific

024270-500MG
101817-762EA 306.15 USD
101817-762
2-[2-Bromo-4-(sec-butyl)phenoxy]acetic acid
2-[2-Bromo-4-(sec-butyl)phenoxy]acetic acid

Hazard: Irritant

Formula: C₁₂H₁₅BrO₃
MDL Number: MFCD08728066
CAS Number: 920122-78-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR