Tetrahydro-2-furanylmethyl-3-hydroxybenzoate

Supplier: Matrix Scientific

024263-500MG
101817-746EA 306.15 USD
101817-746
Tetrahydro-2-furanylmethyl-3-hydroxybenzoate
Tetrahydro-2-furanylmethyl-3-hydroxybenzoate

Hazard: Irritant

Formula: C₁₂H₁₄O₄
MW: 222.24 g/mol
Melting Pt: 101…102 °C
MDL Number: MFCD08721752
CAS Number: 914452-94-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR