Isopropyl-3-hydroxybenzoate

Supplier: Matrix Scientific

024261-500MG
101817-742EA 198.46 USD
101817-742
Isopropyl-3-hydroxybenzoate
Isopropyl-3-hydroxybenzoate

Hazard: Irritant

Formula: C₁₀H₁₂O₃
MW: 180.21 g/mol
MDL Number: MFCD08712957
CAS Number: 53631-77-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR