Phenethyl-3-hydroxybenzoate

Supplier: Matrix Scientific

024259-500MG
101817-738EA 306.15 USD
101817-738
Phenethyl-3-hydroxybenzoate
Phenethyl-3-hydroxybenzoate

Hazard: Irritant

Formula: C₁₅H₁₄O₃
MW: 242.28 g/mol
MDL Number: MFCD08727984
CAS Number: 99287-97-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR