Tetrahydro-2-furanylmethyl-4-hydroxybenzoate

Supplier: Matrix Scientific

024255-500MG
101817-730EA 306.15 USD
101817-730
Tetrahydro-2-furanylmethyl-4-hydroxybenzoate
Tetrahydro-2-furanylmethyl-4-hydroxybenzoate

Hazard: Irritant

Formula: C₁₂H₁₄O₄
MW: 222.24 g/mol
Melting Pt: 101…102 °C
MDL Number: MFCD08727964
CAS Number: 4470-04-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR