Phenethyl-4-hydroxybenzoate

Supplier: Matrix Scientific

024250-500MG
101817-720EA 306.15 USD
101817-720
Phenethyl-4-hydroxybenzoate
Phenethyl-4-hydroxybenzoate

Hazard: Irritant

Formula: C₁₅H₁₄O₃
MW: 242.28 g/mol
MDL Number: MFCD08559391
CAS Number: 83936-28-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR