3-(2-Ethoxyethoxy)aniline hydrochloride

Supplier: Matrix Scientific

024246-500MG
101817-712EA 180 USD
101817-712
3-(2-Ethoxyethoxy)aniline hydrochloride
3-(2-Ethoxyethoxy)aniline hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₆ClNO₂
MDL Number: MFCD08559363
CAS Number: 1049787-84-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR