4-(Hydroxymethyl)-2,6-dimethylphenol

Supplier: Matrix Scientific

024243-500MG
101817-704EA 306.15 USD
101817-704
4-(Hydroxymethyl)-2,6-dimethylphenol
4-(Hydroxymethyl)-2,6-dimethylphenol

Hazard: Irritant

Formula: C₉H₁₂O₂
MDL Number: MFCD02912490
CAS Number: 4397-14-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR