2-[2-(sec-Butyl)phenoxy]propanoic acid

Supplier: Matrix Scientific

024240-500MG
101817-700EA 180 USD
101817-700
2-[2-(sec-Butyl)phenoxy]propanoic acid
2-[2-(sec-Butyl)phenoxy]propanoic acid

Hazard: Irritant

Formula: C₁₃H₁₈O₃
MDL Number: MFCD08559345
CAS Number: 887029-70-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR