N-Benzyl-N-cyclopentylamine

Supplier: Matrix Scientific

024231-500MG
101817-682EA 306.15 USD
101817-682
N-Benzyl-N-cyclopentylamine
N-Benzyl-N-cyclopentylamine

Hazard: Irritant

Formula: C₁₂H₁₇N
Boiling Pt: 114…116 °C (5 Torr)
Melting Pt: 158…159 °C
MDL Number: MFCD04598198
CAS Number: 15205-23-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR