2-(1-Isobutyl-2-piperazinyl)-1-ethanol dihydrochloride

Supplier: Matrix Scientific

024230-500MG
101817-680EA 306.15 USD
101817-680
2-(1-Isobutyl-2-piperazinyl)-1-ethanol dihydrochloride
2-(1-Isobutyl-2-piperazinyl)-1-ethanol dihydrochloride

Hazard: Irritant

Formula: C₁₀H₂₄Cl₂N₂O
MDL Number: MFCD08689408
CAS Number: 1049750-12-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR