3-Bromo-4-(tetrahydro-2-furanylmethoxy)benzoic acid

Supplier: Matrix Scientific

024229-500MG
101817-678EA 306.15 USD
101817-678
3-Bromo-4-(tetrahydro-2-furanylmethoxy)benzoic acid
3-Bromo-4-(tetrahydro-2-furanylmethoxy)benzoic acid

Hazard: Irritant

Formula: C₁₂H₁₃BrO₄
MDL Number: MFCD08134368
CAS Number: 887029-50-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR