[2-(1-Azepanylmethyl)phenyl]methanol

Supplier: Matrix Scientific

024227-500MG
101817-674EA 306.15 USD
101817-674
[2-(1-Azepanylmethyl)phenyl]methanol
[2-(1-Azepanylmethyl)phenyl]methanol

Hazard: Irritant

Formula: C₁₄H₂₁NO
MDL Number: MFCD03211257
CAS Number: 356539-02-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR