2-Isopropoxyacetohydrazide hydrochloride

Supplier: Matrix Scientific

024225-500MG
101817-670EA 192.31 USD
101817-670
2-Isopropoxyacetohydrazide hydrochloride
2-Isopropoxyacetohydrazide hydrochloride

Hazard: Irritant

Formula: C₅H₁₃ClN₂O₂
MDL Number: MFCD08559349
CAS Number: 898747-67-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR