N-Cyclopentyl-N-propylamine hydrochloride

Supplier: Matrix Scientific

024221-500MG
101817-662EA 203.08 USD
101817-662
N-Cyclopentyl-N-propylamine hydrochloride
N-Cyclopentyl-N-propylamine hydrochloride

Hazard: Irritant

Formula: C₈H₁₈ClN
MDL Number: MFCD08559371
CAS Number: 1049760-26-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR