3-(2-Ethoxyethoxy)benzoic acid

Supplier: Matrix Scientific

024220-500MG
101817-660EA 306.15 USD
101817-660
3-(2-Ethoxyethoxy)benzoic acid
3-(2-Ethoxyethoxy)benzoic acid

Hazard: Irritant

Formula: C₁₁H₁₄O₄
Melting Pt: 77…78 °C
MDL Number: MFCD08559379
CAS Number: 898747-62-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR