2-Ethyl-4-methyl-5-amino-3-methyl-2,4-thiophenedicarboxylate

Supplier: Matrix Scientific

024216-500MG
101817-650EA 306.15 USD
101817-650
2-Ethyl-4-methyl-5-amino-3-methyl-2,4-thiophenedicarboxylate
2-Ethyl-4-methyl-5-amino-3-methyl-2,4-thiophenedicarboxylate

Hazard: Irritant

Formula: C₁₀H₁₃NO₄S
MDL Number: MFCD00790509
CAS Number: 77457-04-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR