Ethyl-5-amino-4-cyano-3-methyl-2-thiophenecarboxylate

Supplier: Matrix Scientific

024214-500MG
101817-646EA 306.15 USD
101817-646
Ethyl-5-amino-4-cyano-3-methyl-2-thiophenecarboxylate
Ethyl-5-amino-4-cyano-3-methyl-2-thiophenecarboxylate

Hazard: Irritant

Formula: C₉H₁₀N₂O₂S
MDL Number: MFCD00463473
CAS Number: 23903-46-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR