4-Methoxy-N-(1-phenylethyl)benzamide

Supplier: Matrix Scientific

024212-500MG
101817-642EA 306.15 USD
101817-642
4-Methoxy-N-(1-phenylethyl)benzamide
4-Methoxy-N-(1-phenylethyl)benzamide

Hazard: Irritant

Formula: C₁₆H₁₇NO₂
MDL Number: MFCD00437019
CAS Number: 68162-84-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR