4-(1-Azepanyl)-1-butanol

Supplier: Matrix Scientific

024211-500MG
101817-640EA 203.08 USD
101817-640
4-(1-Azepanyl)-1-butanol
4-(1-Azepanyl)-1-butanol

Hazard: Irritant

Formula: C₁₀H₂₁NO
MDL Number: MFCD03211285
CAS Number: 114960-98-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR