4-(1-Piperidinyl)-1-butanol hydrochloride

Supplier: Matrix Scientific

024208-500MG
101817-634EA 306.15 USD
101817-634
4-(1-Piperidinyl)-1-butanol hydrochloride
4-(1-Piperidinyl)-1-butanol hydrochloride

Hazard: Irritant

Formula: C₉H₂₀ClNO
MDL Number: MFCD12026512
CAS Number: 109967-02-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR