N-Benzyl(cyclopropyl)methanamine

Supplier: Matrix Scientific

024205-500MG
101817-628EA 306.15 USD
101817-628
N-Benzyl(cyclopropyl)methanamine
N-Benzyl(cyclopropyl)methanamine

Hazard: Irritant

Formula: C₁₁H₁₅N
MDL Number: MFCD03129358
CAS Number: 116373-23-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR