4-Amino-N-ethyl-N-(1-naphthyl)benzamide

Supplier: Matrix Scientific

024193-500MG
101817-602EA 253.85 USD
101817-602
4-Amino-N-ethyl-N-(1-naphthyl)benzamide
4-Amino-N-ethyl-N-(1-naphthyl)benzamide

Hazard: Irritant

Formula: C₁₉H₁₈N₂O
MDL Number: MFCD00036002
CAS Number: 6361-29-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR