[2-(1-Pyrrolidinylmethyl)phenyl]methanol

Supplier: Matrix Scientific

024192-500MG
101817-600EA 223.08 USD
101817-600
[2-(1-Pyrrolidinylmethyl)phenyl]methanol
[2-(1-Pyrrolidinylmethyl)phenyl]methanol

Hazard: Irritant

Formula: C₁₂H₁₇NO
MDL Number: MFCD03109885
CAS Number: 91271-58-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR