3-Phenyl-2-(1H-pyrrol-1-yl)propanoic acid

Supplier: Matrix Scientific

024187-500MG
101817-590EA 306.15 USD
101817-590
3-Phenyl-2-(1H-pyrrol-1-yl)propanoic acid
3-Phenyl-2-(1H-pyrrol-1-yl)propanoic acid

Hazard: Irritant

Formula: C₁₃H₁₃NO₂
MDL Number: MFCD03856011
CAS Number: 46687-57-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR