4-Oxo-4-(1-piperidinyl)butanoic acid

Supplier: Matrix Scientific

024185-500MG
101817-586EA 43.08 USD
101817-586
4-Oxo-4-(1-piperidinyl)butanoic acid
4-Oxo-4-(1-piperidinyl)butanoic acid

Hazard: Irritant

Formula: C₉H₁₅NO₃
MDL Number: MFCD00517730
CAS Number: 4672-17-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR