1-(2-Furoyl)piperazine hydrochloride

Supplier: Matrix Scientific

Synonyms: Furan-2-yl(piperazin-1-yl)methanone hydrochloride

024180-500MG
101817-576EA 15.38 USD
101817-576
1-(2-Furoyl)piperazine hydrochloride
1-(2-Furoyl)piperazine hydrochloride

Hazard: Irritant

Formula: C₉H₁₃ClN₂O₂
MW: 216.6647 g/mol
MDL Number: MFCD00236012
CAS Number: 60548-09-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR