Ethyl-3-bromo-4-hydroxybenzoate

Supplier: Matrix Scientific

Synonyms: Ethyl 3-bromo-4-hydroxybenzoate

024177-500MG
101817-570EA 306.15 USD
101817-570
Ethyl-3-bromo-4-hydroxybenzoate
Ethyl-3-bromo-4-hydroxybenzoate

Hazard: Irritant

Formula: C₉H₉BrO₃
MW: 245.08 g/mol
MDL Number: MFCD06204654
CAS Number: 37470-58-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR