N-(2-Hydroxyphenyl)propanamide

Supplier: Matrix Scientific

024175-500MG
101817-566EA 306.15 USD
101817-566
N-(2-Hydroxyphenyl)propanamide
N-(2-Hydroxyphenyl)propanamide

Hazard: Irritant

Formula: C₉H₁₁NO₂
MDL Number: MFCD00791255
CAS Number: 6963-37-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR