2-(1-Cyclohexen-1-yl)-N-(4-methoxybenzyl)-1-ethanamine hydrochloride

Supplier: Matrix Scientific

024157-500MG
101817-526EA 180 USD
101817-526
2-(1-Cyclohexen-1-yl)-N-(4-methoxybenzyl)-1-ethanamine hydrochloride
2-(1-Cyclohexen-1-yl)-N-(4-methoxybenzyl)-1-ethanamine hydrochloride

Hazard: Irritant

Formula: C₁₆H₂₄ClNO
MDL Number: MFCD03876243
CAS Number: 1048640-95-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR