2-(4,5-Dibromo-2-methoxyphenoxy)acetic acid

Supplier: Matrix Scientific

024156-500MG
101817-524EA 203.08 USD
101817-524
2-(4,5-Dibromo-2-methoxyphenoxy)acetic acid
2-(4,5-Dibromo-2-methoxyphenoxy)acetic acid

Hazard: Irritant

Formula: C₉H₈Br₂O₄
Melting Pt: 162 °C
MDL Number: MFCD03109881
CAS Number: 446278-49-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR