N-(4-Methylbenzyl)-1-phenyl-1-ethanamine hydrochloride

Supplier: Matrix Scientific

024153-500MG
101817-518EA 306.15 USD
101817-518
N-(4-Methylbenzyl)-1-phenyl-1-ethanamine hydrochloride
N-(4-Methylbenzyl)-1-phenyl-1-ethanamine hydrochloride

Hazard: Irritant

Formula: C₁₆H₂₀ClN
MDL Number: MFCD03930964
CAS Number: 202199-08-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR