N-[2-(1-Cyclohexen-1-yl)ethyl]-1-propanamine hydrochloride

Supplier: Matrix Scientific

024149-500MG
101817-510EA 306.15 USD
101817-510
N-[2-(1-Cyclohexen-1-yl)ethyl]-1-propanamine hydrochloride
N-[2-(1-Cyclohexen-1-yl)ethyl]-1-propanamine hydrochloride

Hazard: Irritant

Formula: C₁₁H₂₂ClN
MDL Number: MFCD03930961
CAS Number: 356532-69-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR