2-(1-Cyclohexen-1-yl)-N-(3-methylbenzyl)-1-ethanamine hydrochloride

Supplier: Matrix Scientific

024146-500MG
101817-504EA 306.15 USD
101817-504
2-(1-Cyclohexen-1-yl)-N-(3-methylbenzyl)-1-ethanamine hydrochloride
2-(1-Cyclohexen-1-yl)-N-(3-methylbenzyl)-1-ethanamine hydrochloride

Hazard: Irritant

Formula: C₁₆H₂₄ClN
MDL Number: MFCD03930951
CAS Number: 1048640-63-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR