(4-Methoxyphenyl)-N-(tetrahydro-2-furanylmethyl)methanamine hydrochloride

Supplier: Matrix Scientific

024144-500MG
101817-500EA 180 USD
101817-500
(4-Methoxyphenyl)-N-(tetrahydro-2-furanylmethyl)methanamine hydrochloride
(4-Methoxyphenyl)-N-(tetrahydro-2-furanylmethyl)methanamine hydrochloride

Hazard: Irritant

Formula: C₁₃H₂₀ClNO₂
MDL Number: MFCD03930962
CAS Number: 1048640-87-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR