N-(Cyclohexylmethyl)(phenyl)-methanamine hydrochloride

Supplier: Matrix Scientific

024141-500MG
101817-494EA 180 USD
101817-494
N-(Cyclohexylmethyl)(phenyl)-methanamine hydrochloride
N-(Cyclohexylmethyl)(phenyl)-methanamine hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₂ClN
MDL Number: MFCD00760309
CAS Number: 126541-72-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR