N-Benzyl-N-(2-cyclohex-1-en-1-ylethyl)amine hydrochloride

Supplier: Matrix Scientific

024138-500MG
101817-488EA 495.38 USD
101817-488
N-Benzyl-N-(2-cyclohex-1-en-1-ylethyl)amine hydrochloride
N-Benzyl-N-(2-cyclohex-1-en-1-ylethyl)amine hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₂ClN
MW: 251.8 g/mol
MDL Number: MFCD03930931
CAS Number: 118647-00-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR