(1-Naphthoxy)acetic acid

Supplier: Matrix Scientific

Synonyms: 2-(Naphthalen-1-yloxy)acetic acid

024137-500MG
101817-486EA 15.38 USD
101817-486
(1-Naphthoxy)acetic acid
(1-Naphthoxy)acetic acid

Hazard: Irritant

Formula: C₁₂H₁₀O₃
MW: 202.21 g/mol
MDL Number: MFCD00003927
CAS Number: 2976-75-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR