4-(3-Methoxyanilino)-4-oxobutanoic acid

Supplier: Matrix Scientific

024127-500MG
101817-466EA 180 USD
101817-466
4-(3-Methoxyanilino)-4-oxobutanoic acid
4-(3-Methoxyanilino)-4-oxobutanoic acid

Hazard: Irritant

Formula: C₁₁H₁₃NO₄
MDL Number: MFCD00239114
CAS Number: 16141-44-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR