2-(1-Pyrrolidinylcarbonyl)benzoic acid

Supplier: Matrix Scientific

024126-500MG
101817-464EA 180 USD
101817-464
2-(1-Pyrrolidinylcarbonyl)benzoic acid
2-(1-Pyrrolidinylcarbonyl)benzoic acid

Hazard: Irritant

Formula: C₁₂H₁₃NO₃
MDL Number: MFCD01961578
CAS Number: 20320-43-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR