3-Bromo-4-ethoxybenzoic acid

Supplier: Matrix Scientific

024124-500MG
101817-460EA 306.15 USD
101817-460
3-Bromo-4-ethoxybenzoic acid
3-Bromo-4-ethoxybenzoic acid

Hazard: Irritant

Formula: C₉H₉BrO₃
MDL Number: MFCD01143606
CAS Number: 24507-29-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR