2-Isopropoxybenzoic acid

Supplier: Matrix Scientific

024115-1G
101817-442EA 484.62 USD
101817-442
2-Isopropoxybenzoic acid
2-Isopropoxybenzoic acid

Hazard: Irritant

Formula: C₁₀H₁₂O₃
MDL Number: MFCD00156986
CAS Number: 63635-26-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR