5-Bromo-2-(hexyloxy)benzoic acid

Supplier: Matrix Scientific

024102-500MG
101817-416EA 306.15 USD
101817-416
5-Bromo-2-(hexyloxy)benzoic acid
5-Bromo-2-(hexyloxy)benzoic acid

Hazard: Irritant

Formula: C₁₃H₁₇BrO₃
MDL Number: MFCD02668713
CAS Number: 62176-19-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR