5-[2-(2-Ethoxy-2-oxoethyl)-3-oxo-1-piperazinyl]-5-oxopentanoic acid

Supplier: Matrix Scientific

024096-500MG
101817-404EA 306.15 USD
101817-404
5-[2-(2-Ethoxy-2-oxoethyl)-3-oxo-1-piperazinyl]-5-oxopentanoic acid
5-[2-(2-Ethoxy-2-oxoethyl)-3-oxo-1-piperazinyl]-5-oxopentanoic acid

Hazard: Irritant

Formula: C₁₃H₂₀N₂O₆
MDL Number: MFCD05881340
CAS Number: 1025063-22-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR