2-[(3-Bromo[1,1'-biphenyl]-4-yl)oxy]acetic acid

Supplier: Matrix Scientific

024094-500MG
101817-400EA 306.15 USD
101817-400
2-[(3-Bromo[1,1'-biphenyl]-4-yl)oxy]acetic acid
2-[(3-Bromo[1,1'-biphenyl]-4-yl)oxy]acetic acid

Hazard: Irritant

Formula: C₁₄H₁₁BrO₃
MDL Number: MFCD06662559
CAS Number: 625820-74-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR